Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTGTGGAAGGTGTTCACGTAG[A/G]CTGCGCCTCCCAGGAGCCCCTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607042 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLN3 - CLN3, battenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000086.2 | 1335 | Missense Mutation | GCC,GTC | A,V 388 | NP_000077.1 | |
NM_001042432.1 | 1335 | Missense Mutation | GCC,GTC | A,V 388 | NP_001035897.1 | |
NM_001286104.1 | 1335 | Missense Mutation | GCC,GTC | A,V 364 | NP_001273033.1 | |
NM_001286105.1 | 1335 | Missense Mutation | GCC,GTC | A,V 288 | NP_001273034.1 | |
NM_001286109.1 | 1335 | Missense Mutation | GCC,GTC | A,V 310 | NP_001273038.1 | |
NM_001286110.1 | 1335 | Missense Mutation | GCC,GTC | A,V 334 | NP_001273039.1 |