Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCTGAGAGGAGTGATTGCGTT[C/T]CACATGTTGACTGAGAAATTTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM7 - PR domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098173.1 | 1227 | Nonsense Mutation | TGA,TGG | *,W 437 | NP_001091643.1 | |
XM_005256274.3 | 1227 | Nonsense Mutation | TGA,TGG | *,W 437 | XP_005256331.1 | |
XM_011522829.1 | 1227 | Missense Mutation | GAA,GGA | E,G 343 | XP_011521131.1 | |
XM_011522831.2 | 1227 | Intron | XP_011521133.1 | |||
XM_017022882.1 | 1227 | Intron | XP_016878371.1 | |||
XM_017022883.1 | 1227 | Intron | XP_016878372.1 | |||
XM_017022884.1 | 1227 | Intron | XP_016878373.1 |