Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTGGGAGCGAGTGGCCCGGA[C/T]GCTGGAGGCCTGGGAGTGGGGAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613190 MIM: 604905 | ||||||||||||||||||||
Literature Links: |
DNAAF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAAF1 - dynein axonemal assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318756.1 | 2784 | Intron | NP_001305685.1 | |||
NM_178452.5 | 2784 | Intron | NP_848547.4 | |||
XM_006721129.2 | 2784 | Intron | XP_006721192.1 | |||
XM_011522853.2 | 2784 | Intron | XP_011521155.1 | |||
XM_011522854.2 | 2784 | Intron | XP_011521156.1 | |||
XM_011522855.2 | 2784 | Intron | XP_011521157.1 | |||
XM_011522857.2 | 2784 | Intron | XP_011521159.1 | |||
XM_011522858.2 | 2784 | Intron | XP_011521160.1 | |||
XM_017022918.1 | 2784 | Intron | XP_016878407.1 | |||
XM_017022919.1 | 2784 | Intron | XP_016878408.1 | |||
XM_017022920.1 | 2784 | Intron | XP_016878409.1 | |||
XM_017022921.1 | 2784 | Intron | XP_016878410.1 | |||
XM_017022922.1 | 2784 | Intron | XP_016878411.1 |
TAF1C - TATA-box binding protein associated factor, RNA polymerase I subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243156.1 | 2784 | Missense Mutation | NP_001230085.1 | |||
NM_001243157.1 | 2784 | Missense Mutation | NP_001230086.1 | |||
NM_001243158.1 | 2784 | Missense Mutation | NP_001230087.1 | |||
NM_001243159.1 | 2784 | Missense Mutation | NP_001230088.1 | |||
NM_001243160.1 | 2784 | Missense Mutation | NP_001230089.1 | |||
NM_005679.3 | 2784 | Missense Mutation | NP_005670.3 | |||
NM_139353.2 | 2784 | Missense Mutation | NP_647610.2 | |||
XM_005256226.3 | 2784 | Missense Mutation | XP_005256283.1 | |||
XM_005256227.3 | 2784 | Missense Mutation | XP_005256284.1 | |||
XM_006721325.3 | 2784 | Missense Mutation | XP_006721388.1 | |||
XM_006721326.3 | 2784 | Missense Mutation | XP_006721389.1 | |||
XM_017023845.1 | 2784 | Missense Mutation | XP_016879334.1 | |||
XM_017023846.1 | 2784 | Missense Mutation | XP_016879335.1 | |||
XM_017023847.1 | 2784 | Missense Mutation | XP_016879336.1 | |||
XM_017023848.1 | 2784 | Missense Mutation | XP_016879337.1 |