Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGATAGTCAGAGGAGCTGTGT[G/T]CTTCCCTCCATCACGTGTCCCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608107 MIM: 603231 | ||||||||||||||||||||
Literature Links: |
MEFV PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEFV - Mediterranean fever | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000243.2 | 2474 | Missense Mutation | AAC,CAC | N,H 766 | NP_000234.1 | |
NM_001198536.1 | 2474 | UTR 3 | NP_001185465.1 | |||
XM_017023236.1 | 2474 | Missense Mutation | AAC,CAC | N,H 765 | XP_016878725.1 |
ZNF200 - zinc finger protein 200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |