Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCATATCCTGGGGCAACCCCTT[C/T]CAGCCCAAGGTGCCGCACACTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605584 | ||||||||||||||||||||
Literature Links: |
DHX38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX38 - DEAH-box helicase 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMFBP1 - polyamine modulated factor 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160213.1 | 3019 | Missense Mutation | NP_001153685.1 | |||
NM_031293.2 | 3019 | Missense Mutation | NP_112583.2 | |||
XM_011523357.2 | 3019 | Nonsense Mutation | XP_011521659.1 | |||
XM_011523358.2 | 3019 | Nonsense Mutation | XP_011521660.1 | |||
XM_011523360.2 | 3019 | Nonsense Mutation | XP_011521662.1 | |||
XM_011523361.2 | 3019 | Nonsense Mutation | XP_011521663.1 | |||
XM_011523362.2 | 3019 | Nonsense Mutation | XP_011521664.1 |