Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAGGCCAGTTGAGGTGCGGAA[C/G]GAGGAAATCACCTTAAAGCAGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ADGRG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRG3 - adhesion G protein-coupled receptor G3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DRC7 - dynein regulatory complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289162.1 | 206 | Missense Mutation | AAC,AAG | N,K 39 | NP_001276091.1 | |
NM_001289163.1 | 206 | Missense Mutation | AAC,AAG | N,K 39 | NP_001276092.1 | |
NM_032269.5 | 206 | Missense Mutation | AAC,AAG | N,K 39 | NP_115645.4 | |
XM_011523377.1 | 206 | Missense Mutation | AAC,AAG | N,K 39 | XP_011521679.1 | |
XM_011523378.1 | 206 | Missense Mutation | AAC,AAG | N,K 39 | XP_011521680.1 |