Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCACTACAGAATAGAGAACCCA[A/G]ATTTTTATATACAAAGTGCTTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITFG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITFG1 - integrin alpha FG-GAP repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305002.1 | 1933 | UTR 3 | NP_001291931.1 | |||
NM_030790.4 | 1933 | UTR 3 | NP_110417.2 |
ITFG1-AS1 - ITFG1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |