Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTTTCTGCATTTAGATAAAAA[C/T]TCTTGGCTAATCTGTAAGAAGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOPNL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOPNL - FGFR1OP N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304497.1 | 1647 | UTR 3 | NP_001291426.1 | |||
NM_001304498.1 | 1647 | UTR 3 | NP_001291427.1 | |||
NM_001304499.1 | 1647 | UTR 3 | NP_001291428.1 | |||
NM_001304500.1 | 1647 | UTR 3 | NP_001291429.1 | |||
NM_001304502.1 | 1647 | UTR 3 | NP_001291431.1 | |||
NM_144600.3 | 1647 | UTR 3 | NP_653201.1 |
MYH11 - myosin heavy chain 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |