Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAACCTCTGATAGTGATCAGGAT[G/T]GGTCACAGAGGAGCCGACTGGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZP2 - zona pellucida glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290104.1 | 2212 | Missense Mutation | AAT,CAT | N,H 577 | NP_001277033.1 | |
NM_003460.2 | 2212 | Missense Mutation | AAT,CAT | N,H 586 | NP_003451.1 | |
XM_011545942.2 | 2212 | Missense Mutation | AAT,CAT | N,H 586 | XP_011544244.1 |