Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610886 MIM: 605431 MIM: 611994 MIM: 601817 MIM: 611659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EME2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EME2 - essential meiotic structure-specific endonuclease subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP3 - mitogen-activated protein kinase 8 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS34 - mitochondrial ribosomal protein S34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300900.1 | 833 | UTR 3 | NP_001287829.1 | |||
NM_023936.1 | 833 | UTR 3 | NP_076425.1 | |||
XM_017023595.1 | 833 | UTR 3 | XP_016879084.1 |
NME3 - NME/NM23 nucleoside diphosphate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002513.2 | 833 | Intron | NP_002504.2 | |||
XM_005255332.4 | 833 | Intron | XP_005255389.1 | |||
XM_011522503.2 | 833 | Intron | XP_011520805.1 | |||
XM_011522504.2 | 833 | Intron | XP_011520806.1 |
SPSB3 - splA/ryanodine receptor domain and SOCS box containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |