Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGCAACCTGGGGTGATCCCG[C/T]GGGCTCTCATGGACCTCCTGCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603213 MIM: 600999 | ||||||||||||||||||||
Literature Links: |
KIF22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF22 - kinesin family member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256269.1 | 629 | Missense Mutation | CGG,TGG | R,W 81 | NP_001243198.1 | |
NM_001256270.1 | 629 | Missense Mutation | CGG,TGG | R,W 81 | NP_001243199.1 | |
NM_007317.2 | 629 | Missense Mutation | CGG,TGG | R,W 149 | NP_015556.1 |
MAZ - MYC associated zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |