Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGTCCCTGATGTCCCACCAGA[C/T]CCTGGAGCACATGGAGGGAGCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDP2 - pyruvate dehyrogenase phosphatase catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256064.4 | 835 | Missense Mutation | ACC,ATC | T,I 169 | XP_005256121.1 | |
XM_005256066.4 | 835 | Missense Mutation | ACC,ATC | T,I 169 | XP_005256123.1 | |
XM_005256067.4 | 835 | Missense Mutation | ACC,ATC | T,I 169 | XP_005256124.1 | |
XM_005256068.4 | 835 | Missense Mutation | ACC,ATC | T,I 169 | XP_005256125.1 | |
XM_017023506.1 | 835 | Missense Mutation | ACC,ATC | T,I 169 | XP_016878995.1 |