Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGTCCATTGCACGTAGGAGAAC[A/G]ACTGTCCATTCCCGGGACAAGCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMC7 - transmembrane channel like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160364.2 | 390 | UTR 5 | NP_001153836.1 | |||
NM_001300732.1 | 390 | Silent Mutation | ACA,ACG | T,T 54 | NP_001287661.1 | |
NM_001324263.1 | 390 | UTR 5 | NP_001311192.1 | |||
NM_001324265.1 | 390 | Silent Mutation | ACA,ACG | T,T 54 | NP_001311194.1 | |
NM_001324268.1 | 390 | UTR 5 | NP_001311197.1 | |||
NM_024847.4 | 390 | Silent Mutation | ACA,ACG | T,T 54 | NP_079123.3 |