Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGGGAACACCTGGAACCAGCT[C/G]AGAAGAATTTGTATCAGGATGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604752 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF267 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF267 - zinc finger protein 267 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265588.1 | 309 | UTR 5 | NP_001252517.1 | |||
NM_003414.5 | 309 | Missense Mutation | CAG,GAG | Q,E 25 | NP_003405.3 | |
XM_017022865.1 | 309 | UTR 5 | XP_016878354.1 | |||
XM_017022866.1 | 309 | UTR 5 | XP_016878355.1 |