Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGGAGATGGGCAGAAGGCCCC[A/G]AGCTTCCAGTGGCGGAACTACAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609085 MIM: 610930 MIM: 611052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL19 - F-box and leucine rich repeat protein 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORAI3 - ORAI calcium release-activated calcium modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014712.2 | 130 | Silent Mutation | CCA,CCG | P,P 13 | NP_055527.1 | |
XM_005255723.1 | 130 | Silent Mutation | CCA,CCG | P,P 13 | XP_005255780.1 | |
XM_006721106.3 | 130 | Silent Mutation | CCA,CCG | P,P 13 | XP_006721169.1 | |
XM_017023909.1 | 130 | Silent Mutation | CCA,CCG | P,P 13 | XP_016879398.1 |