Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCTTGCCCAGGCCCTGGAAGA[C/G]GCGGTGGGGCAGGTGCGTGAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601489 MIM: 610779 MIM: 611659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFALS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFALS - insulin like growth factor binding protein acid labile subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146006.1 | 1552 | Missense Mutation | CTC,GTC | L,V 491 | NP_001139478.1 | |
NM_004970.2 | 1552 | Missense Mutation | CTC,GTC | L,V 453 | NP_004961.1 |
NUBP2 - nucleotide binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPSB3 - splA/ryanodine receptor domain and SOCS box containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |