Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCTTGATACTTACGGTCCCACC[A/G]TTGAGCACGACGGCCATCTCAGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605948 | ||||||||||||||||||||
Literature Links: |
GPRC5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPRC5B - G protein-coupled receptor class C group 5 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304771.1 | 1344 | Silent Mutation | AAC,AAT | N,N 517 | NP_001291700.1 | |
NM_016235.2 | 1344 | Silent Mutation | AAC,AAT | N,N 386 | NP_057319.1 | |
XM_006721051.2 | 1344 | Silent Mutation | AAC,AAT | N,N 386 | XP_006721114.1 | |
XM_006721052.2 | 1344 | Silent Mutation | AAC,AAT | N,N 386 | XP_006721115.1 |
IQCK - IQ motif containing K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |