Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTTCTTGGCGCGGTCTGCTGC[A/G]TCCAGGGCAGCCATGTTGCGGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605893 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDIPT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDIPT - CDP-diacylglycerol--inositol 3-phosphatidyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286585.1 | 911 | Silent Mutation | NP_001273514.1 | |||
NM_001286586.1 | 911 | Silent Mutation | NP_001273515.1 | |||
NM_006319.4 | 911 | Silent Mutation | NP_006310.1 | |||
XM_005255038.4 | 911 | Missense Mutation | XP_005255095.1 |
CDIPT-AS1 - CDIPT antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |