Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCAACCCTCAGTCTCTCTGCC[A/G]TTTCTTGATTCCTTTCCAAACCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604009 MIM: 607838 | ||||||||||||||||||||
Literature Links: |
BAIAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAIAP3 - BAI1 associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199096.1 | 993 | Intron | NP_001186025.1 | |||
NM_001199097.1 | 993 | Intron | NP_001186026.1 | |||
NM_001199098.1 | 993 | Intron | NP_001186027.1 | |||
NM_001199099.1 | 993 | Intron | NP_001186028.1 | |||
NM_001286464.1 | 993 | Intron | NP_001273393.1 | |||
NM_003933.4 | 993 | Intron | NP_003924.2 | |||
XM_011522728.1 | 993 | Intron | XP_011521030.1 | |||
XM_011522729.1 | 993 | Intron | XP_011521031.1 | |||
XM_011522730.2 | 993 | Intron | XP_011521032.1 |
GNPTG - N-acetylglucosamine-1-phosphate transferase gamma subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSR3 - TSR3, acp transferase ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001410.2 | 993 | Missense Mutation | CGG,TGG | R,W 309 | NP_001001410.1 |