Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTATACCATTTTCATTTAATTA[C/T]GTAATTTTTGAATGTTTGAGTTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IST1 - IST1, ESCRT-III associated factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270975.1 | Intron | NP_001257904.1 | ||||
NM_001270976.1 | Intron | NP_001257905.1 | ||||
NM_001270977.1 | Intron | NP_001257906.1 | ||||
NM_001270978.1 | Intron | NP_001257907.1 | ||||
NM_001270979.1 | Intron | NP_001257908.1 | ||||
NM_014761.3 | Intron | NP_055576.2 |