Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTTCTTGGCATTTGTTCTCTGC[C/T]GGTCCTGGAAGAAGTGGGGAAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603603 | ||||||||||||||||||||
Literature Links: |
BFAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BFAR - bifunctional apoptosis regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G10 - phospholipase A2 group X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003561.2 | 704 | Silent Mutation | CCA,CCG | P,P 120 | NP_003552.1 | |
XM_011522695.2 | 704 | Intron | XP_011520997.1 | |||
XM_011522696.2 | 704 | Intron | XP_011520998.1 | |||
XM_011522697.1 | 704 | Intron | XP_011520999.1 | |||
XM_011522698.1 | 704 | Intron | XP_011521000.1 |