Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCCTGAGGAGCCAGCTCCCGCC[A/G]TCCAGGAGGACGAGGCTGAGAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H18 - zinc finger CCCH-type containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294340.1 | 624 | Missense Mutation | ATC,GTC | I,V 142 | NP_001281269.1 | |
NM_144604.3 | 624 | Missense Mutation | ATC,GTC | I,V 142 | NP_653205.3 | |
XM_011522863.1 | 624 | Missense Mutation | ATC,GTC | I,V 142 | XP_011521165.1 | |
XM_011522865.1 | 624 | Intron | XP_011521167.1 | |||
XM_017022934.1 | 624 | Missense Mutation | ATC,GTC | I,V 142 | XP_016878423.1 |