Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGGACAAGTTCCTGACTGGTGT[A/G]GCCGTGGTGCTGACCGAAAAATACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 141800 MIM: 141850 MIM: 609639 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HBA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HBA1 - hemoglobin subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBA2 - hemoglobin subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBM - hemoglobin subunit mu | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003938.3 | 420 | Silent Mutation | GTA,GTG | V,V 132 | NP_001003938.1 |