Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGGCAGGGGCCTGGGTCTCCA[C/T]GGGGGCGGCAGGCAGGAACACGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616957 | ||||||||||||||||||||
Literature Links: |
TPPP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPPP3 - tubulin polymerization promoting protein family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC1 - zinc finger DHHC-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323627.1 | 744 | Missense Mutation | ATG,GTG | M,V 236 | NP_001310556.1 | |
NM_013304.2 | 744 | Missense Mutation | ATG,GTG | M,V 236 | NP_037436.1 | |
XM_005255915.1 | 744 | Missense Mutation | ATG,GTG | M,V 236 | XP_005255972.1 | |
XM_006721185.1 | 744 | Missense Mutation | ATG,GTG | M,V 236 | XP_006721248.1 | |
XM_011523058.1 | 744 | Missense Mutation | ATG,GTG | M,V 236 | XP_011521360.1 | |
XM_011523059.1 | 744 | Missense Mutation | ATG,GTG | M,V 236 | XP_011521361.1 | |
XM_011523060.1 | 744 | Missense Mutation | ATG,GTG | M,V 151 | XP_011521362.1 |