Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGCACGGTACTCACTGTAACTT[C/G]AGCATAATCTGTTGGCATGTGAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZC3H7A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZC3H7A - zinc finger CCCH-type containing 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014153.3 | 2516 | Missense Mutation | CAA,GAA | Q,E 852 | NP_054872.2 | |
XM_006720877.2 | 2516 | Missense Mutation | CAA,GAA | Q,E 852 | XP_006720940.1 | |
XM_011522463.1 | 2516 | Missense Mutation | CAA,GAA | Q,E 852 | XP_011520765.1 | |
XM_011522464.2 | 2516 | Missense Mutation | CAA,GAA | Q,E 613 | XP_011520766.1 | |
XM_017023174.1 | 2516 | Missense Mutation | CAA,GAA | Q,E 751 | XP_016878663.1 | |
XM_017023175.1 | 2516 | Missense Mutation | CAA,GAA | Q,E 461 | XP_016878664.1 |