Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGCGGTTCTTACAGAAGCTCC[A/G]GCATCCCAACACCATTCAGTACCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987231 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987231 - uncharacterized LOC107987231 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAOK2 - TAO kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252043.1 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | NP_001238972.1 | |
NM_004783.3 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | NP_004774.1 | |
NM_016151.3 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | NP_057235.2 | |
XM_011545982.1 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | XP_011544284.1 | |
XM_011545983.2 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | XP_011544285.1 | |
XM_011545984.1 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | XP_011544286.1 | |
XM_011545985.1 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | XP_011544287.1 | |
XM_011545986.2 | 1068 | Missense Mutation | CAG,CGG | Q,R 84 | XP_011544288.1 |
TMEM219 - transmembrane protein 219 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |