Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAAATTACAAATTGTTTGGTGA[C/T]TATTTTGATGACCTCTTGAGCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603385 | ||||||||||||||||||||
Literature Links: |
NAE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAE1 - NEDD8 activating enzyme E1 subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018159.1 | 1414 | Missense Mutation | ATC,GTC | I,V 503 | NP_001018169.1 | |
NM_001018160.1 | 1414 | Missense Mutation | ATC,GTC | I,V 420 | NP_001018170.1 | |
NM_001286500.1 | 1414 | Missense Mutation | ATC,GTC | I,V 512 | NP_001273429.1 | |
NM_003905.3 | 1414 | Missense Mutation | ATC,GTC | I,V 509 | NP_003896.1 | |
XM_005256215.1 | 1414 | Missense Mutation | ATC,GTC | I,V 420 | XP_005256272.1 | |
XM_011523422.2 | 1414 | Missense Mutation | ATC,GTC | I,V 420 | XP_011521724.1 |
TERB1 - telomere repeat binding bouquet formation protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136505.1 | 1414 | Intron | NP_001129977.1 | |||
XM_011523004.2 | 1414 | Intron | XP_011521306.1 | |||
XM_011523005.2 | 1414 | Intron | XP_011521307.1 | |||
XM_011523006.2 | 1414 | Intron | XP_011521308.1 | |||
XM_011523007.2 | 1414 | Intron | XP_011521309.1 | |||
XM_011523008.2 | 1414 | Intron | XP_011521310.1 | |||
XM_011523009.2 | 1414 | Intron | XP_011521311.1 | |||
XM_011523010.2 | 1414 | Intron | XP_011521312.1 | |||
XM_011523011.2 | 1414 | Intron | XP_011521313.1 | |||
XM_011523012.2 | 1414 | Intron | XP_011521314.1 | |||
XM_011523014.2 | 1414 | Intron | XP_011521316.1 | |||
XM_011523015.2 | 1414 | Intron | XP_011521317.1 | |||
XM_011523016.2 | 1414 | Intron | XP_011521318.1 | |||
XM_011523017.1 | 1414 | Intron | XP_011521319.1 | |||
XM_011523018.1 | 1414 | Intron | XP_011521320.1 | |||
XM_011523020.1 | 1414 | Intron | XP_011521322.1 | |||
XM_017023157.1 | 1414 | Intron | XP_016878646.1 | |||
XM_017023158.1 | 1414 | Intron | XP_016878647.1 | |||
XM_017023159.1 | 1414 | Intron | XP_016878648.1 |