Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATATCAGTCGGGCAGCTGTGGAT[C/T]GAATAATCCGGGTGGATCATGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601683 | ||||||||||||||||||||
Literature Links: |
COQ7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COQ7 - coenzyme Q7, hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190983.1 | 170 | Nonsense Mutation | CGA,TGA | R,* 13 | NP_001177912.1 | |
NM_016138.4 | 170 | Nonsense Mutation | CGA,TGA | R,* 51 | NP_057222.2 |
LOC102723385 - uncharacterized LOC102723385 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMC7 - transmembrane channel like 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |