Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAGTTTGTCTTCTCTTTCAAC[G/T]CCATGGCGTCCTCTTCCATGCACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614975 MIM: 601089 | ||||||||||||||||||||
Literature Links: |
FENDRR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FENDRR - FOXF1 adjacent non-coding developmental regulatory RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOXF1 - forkhead box F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001451.2 | 1091 | Missense Mutation | GCC,TCC | A,S 350 | NP_001442.2 |