Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGGAGCCTGGGAGGCCCACAC[C/T]CACCTACCATCTTGTCCCTAACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605641 | ||||||||||||||||||||
Literature Links: |
SLC7A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC7A6 - solute carrier family 7 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076785.2 | 815 | Missense Mutation | CCC,CTC | P,L 11 | NP_001070253.1 | |
NM_003983.5 | 815 | Missense Mutation | CCC,CTC | P,L 11 | NP_003974.3 | |
XM_011523433.1 | 815 | Missense Mutation | CCC,CTC | P,L 11 | XP_011521735.1 | |
XM_011523434.1 | 815 | Missense Mutation | CCC,CTC | P,L 11 | XP_011521736.1 | |
XM_011523438.1 | 815 | Missense Mutation | CCC,CTC | P,L 11 | XP_011521740.1 | |
XM_011523439.1 | 815 | UTR 5 | XP_011521741.1 | |||
XM_017023851.1 | 815 | Intron | XP_016879340.1 |