Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCGCACGGCCCGCATGGTGGTC[A/G]CCGGGTCCGTCTCACCTGCGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAGH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAGH - hydroxyacylglutathione hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040427.1 | 905 | Missense Mutation | GCG,GTG | A,V 242 | NP_001035517.1 | |
NM_001286249.1 | 905 | UTR 3 | NP_001273178.1 | |||
NM_005326.4 | 905 | Missense Mutation | GCG,GTG | A,V 290 | NP_005317.2 | |
XM_011522469.2 | 905 | UTR 3 | XP_011520771.1 | |||
XM_011522470.2 | 905 | UTR 3 | XP_011520772.1 | |||
XM_017023195.1 | 905 | UTR 3 | XP_016878684.1 |