Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAACCAAAATAGAAGCGATACC[A/G]AGTATCTTACGGTCTGCAGGACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616663 MIM: 616183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORD118 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORD118 - small nucleolar RNA, C/D box 118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM107 - transmembrane protein 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032354.3 | 838 | UTR 3 | NP_115730.2 | |||
NM_183065.2 | 838 | UTR 3 | NP_898888.1 |