Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCTCAGCCCCAGACTCACCCAC[A/G]CCCCCGAAGGGCAGAGAGTGCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100660 | ||||||||||||||||||||
Literature Links: |
ALDH3A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH3A1 - aldehyde dehydrogenase 3 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000691.4 | 1156 | Silent Mutation | GGC,GGT | G,G 404 | NP_000682.3 | |
NM_001135167.1 | 1156 | Silent Mutation | GGC,GGT | G,G 404 | NP_001128639.1 | |
NM_001135168.1 | 1156 | Silent Mutation | GGC,GGT | G,G 404 | NP_001128640.1 | |
XM_005256522.1 | 1156 | Silent Mutation | GGC,GGT | G,G 521 | XP_005256579.1 | |
XM_005256523.2 | 1156 | Silent Mutation | GGC,GGT | G,G 521 | XP_005256580.1 | |
XM_005256524.3 | 1156 | Silent Mutation | GGC,GGT | G,G 521 | XP_005256581.1 | |
XM_011523730.1 | 1156 | Silent Mutation | GGC,GGT | G,G 331 | XP_011522032.1 | |
XM_011523731.1 | 1156 | Silent Mutation | GGC,GGT | G,G 331 | XP_011522033.1 |