Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACGCCGTGCTGGCCACCGGCT[C/T]GGGGATCGTCATCATTCGCTCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611627 | ||||||||||||||||||||
Literature Links: |
FAM57A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM57A - family with sequence similarity 57 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318006.1 | 263 | Missense Mutation | TCG,TTG | S,L 54 | NP_001304935.1 | |
NM_001318007.1 | 263 | Missense Mutation | TCG,TTG | S,L 54 | NP_001304936.1 | |
NM_001318008.1 | 263 | Missense Mutation | TCG,TTG | S,L 54 | NP_001304937.1 | |
NM_024792.2 | 263 | Missense Mutation | TCG,TTG | S,L 54 | NP_079068.1 |