Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCAGGTCTATGTCCCGGTTTCC[C/T]GCAGTCGCGGGCAGGGCGCCAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607570 | ||||||||||||||||||||
Literature Links: |
DHX40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX40 - DEAH-box helicase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166301.1 | 108 | Silent Mutation | CCC,CCT | P,P 5 | NP_001159773.1 | |
NM_024612.4 | 108 | Silent Mutation | CCC,CCT | P,P 5 | NP_078888.4 | |
XM_006722083.2 | 108 | Silent Mutation | CCC,CCT | P,P 5 | XP_006722146.1 | |
XM_011525253.2 | 108 | Silent Mutation | CCC,CCT | P,P 5 | XP_011523555.1 | |
XM_011525254.2 | 108 | UTR 5 | XP_011523556.1 | |||
XM_017025080.1 | 108 | Silent Mutation | CCC,CCT | P,P 5 | XP_016880569.1 | |
XM_017025081.1 | 108 | UTR 5 | XP_016880570.1 | |||
XM_017025082.1 | 108 | UTR 5 | XP_016880571.1 | |||
XM_017025083.1 | 108 | UTR 5 | XP_016880572.1 |