Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGGTGTCTGCCACATAGGTCA[C/T]TATTTGCTCCAAAGTAAAGTTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614955 MIM: 614957 | ||||||||||||||||||||
Literature Links: |
SLFN12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLFN12 - schlafen family member 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLFN13 - schlafen family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144682.5 | 2641 | Missense Mutation | NP_653283.3 | |||
XM_005257922.2 | 2641 | Missense Mutation | XP_005257979.1 | |||
XM_011524383.2 | 2641 | Missense Mutation | XP_011522685.1 | |||
XM_017024232.1 | 2641 | Missense Mutation | XP_016879721.1 | |||
XM_017024233.1 | 2641 | Missense Mutation | XP_016879722.1 |