Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGAGAACTTACAGAATGCCCT[A/G]TACTTCGATATGGTGCCAGAGTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603330 MIM: 194524 | ||||||||||||||||||||
Literature Links: |
DNAH9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH9 - dynein axonemal heavy chain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001372.3 | 12851 | Silent Mutation | CTA,CTG | L,L 4297 | NP_001363.2 | |
NM_004662.2 | 12851 | Silent Mutation | CTA,CTG | L,L 609 | NP_004653.2 | |
XM_011523703.1 | 12851 | Silent Mutation | CTA,CTG | L,L 4312 | XP_011522005.1 | |
XM_017024292.1 | 12851 | Silent Mutation | CTA,CTG | L,L 4273 | XP_016879781.1 | |
XM_017024293.1 | 12851 | Silent Mutation | CTA,CTG | L,L 3606 | XP_016879782.1 | |
XM_017024294.1 | 12851 | Intron | XP_016879783.1 | |||
XM_017024295.1 | 12851 | Intron | XP_016879784.1 |
ZNF18 - zinc finger protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303281.1 | 12851 | Intron | NP_001290210.1 | |||
NM_001303282.1 | 12851 | Intron | NP_001290211.1 | |||
NM_144680.3 | 12851 | Intron | NP_653281.2 | |||
XM_011524002.2 | 12851 | Intron | XP_011522304.1 | |||
XM_017025006.1 | 12851 | Intron | XP_016880495.1 | |||
XM_017025007.1 | 12851 | Intron | XP_016880496.1 | |||
XM_017025008.1 | 12851 | Intron | XP_016880497.1 | |||
XM_017025009.1 | 12851 | Intron | XP_016880498.1 | |||
XM_017025010.1 | 12851 | Intron | XP_016880499.1 | |||
XM_017025011.1 | 12851 | Intron | XP_016880500.1 |