Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGCCACTAGCATTGGTGGTGGC[A/G]CATGGGTTGCAGGGGAGCCTAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602762 MIM: 602763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT33B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT33B - keratin 33B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT34 - keratin 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021013.3 | 1333 | Silent Mutation | NP_066293.2 | |||
XM_011524793.1 | 1333 | Silent Mutation | XP_011523095.1 |