Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAAGATGCAGGTCAAAAGCATG[A/G]TTGCTGTAATATATAAAGTGTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3A2 - aldehyde dehydrogenase 3 family member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000382.2 | Intron | NP_000373.1 | ||||
NM_001031806.1 | Intron | NP_001026976.1 | ||||
XM_011523732.1 | Intron | XP_011522034.1 | ||||
XM_011523733.1 | Intron | XP_011522035.1 | ||||
XM_017024355.1 | Intron | XP_016879844.1 | ||||
XM_017024356.1 | Intron | XP_016879845.1 | ||||
XM_017024357.1 | Intron | XP_016879846.1 | ||||
XM_017024358.1 | Intron | XP_016879847.1 |