Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAGGAGTACCGAGACTGGGTCT[C/T]GGATAACATGTTTTCCAAAGCAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606194 | ||||||||||||||||||||
Literature Links: |
KRT23 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT23 - keratin 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282433.1 | 650 | Missense Mutation | AAG,GAG | K,E 180 | NP_001269362.1 | |
NM_015515.4 | 650 | Missense Mutation | AAG,GAG | K,E 317 | NP_056330.3 | |
XM_005257200.4 | 650 | Missense Mutation | AAG,GAG | K,E 180 | XP_005257257.1 | |
XM_011524595.1 | 650 | Missense Mutation | AAG,GAG | K,E 180 | XP_011522897.1 |