Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATTCTGATAAGATTTCTCTGCT[A/G]TGGAGGAGCCTGACTCACTTTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609086 MIM: 604311 | ||||||||||||||||||||
Literature Links: |
FBXL20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXL20 - F-box and leucine rich repeat protein 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED1 - mediator complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004774.3 | 4557 | Missense Mutation | ACA,ATA | T,I 1471 | NP_004765.2 | |
XM_005257465.3 | 4557 | Missense Mutation | ACA,ATA | T,I 1299 | XP_005257522.1 | |
XM_006721957.1 | 4557 | Intron | XP_006722020.1 | |||
XM_017024779.1 | 4557 | Missense Mutation | ACA,ATA | T,I 1252 | XP_016880268.1 |