Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAAGCCCATGAAAAGCACACAA[A/G]GAGGTTTGAGATACTGGAAGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS13 - dehydrogenase/reductase 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF12 - PHD finger protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033561.1 | 2996 | Intron | NP_001028733.1 | |||
NM_001290131.1 | 2996 | Silent Mutation | TCC,TCT | S,S 822 | NP_001277060.1 | |
NM_020889.2 | 2996 | Intron | NP_065940.1 | |||
XM_005258015.3 | 2996 | Intron | XP_005258072.1 | |||
XM_005258016.3 | 2996 | Intron | XP_005258073.1 | |||
XM_011525078.2 | 2996 | Intron | XP_011523380.1 | |||
XM_011525079.2 | 2996 | Intron | XP_011523381.1 | |||
XM_011525082.2 | 2996 | Intron | XP_011523384.1 | |||
XM_011525083.2 | 2996 | Intron | XP_011523385.1 | |||
XM_017024903.1 | 2996 | Intron | XP_016880392.1 | |||
XM_017024904.1 | 2996 | Intron | XP_016880393.1 |