Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCAGTGCCCCAGCTCCTACCT[C/G]CCTCCCGAATGAGTTTCTTGGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160993 MIM: 608795 | ||||||||||||||||||||
Literature Links: |
MIR6784 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6784 - microRNA 6784 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMT1 - N-myristoyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCD3 - phospholipase C delta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133373.4 | 1832 | Missense Mutation | GCA,GGA | A,G 570 | NP_588614.1 | |
XM_011524253.2 | 1832 | Intron | XP_011522555.1 |