Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGCTGCAGGCGCAGCTCGAGC[A/G]CCTGACGCGCCAGGTGGAGGCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMTNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMTNL2 - smoothelin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114974.1 | 294 | Missense Mutation | CAC,CGC | H,R 76 | NP_001108446.1 | |
NM_198501.2 | 294 | Intron | NP_940903.2 | |||
XM_005256622.3 | 294 | Missense Mutation | CAC,CGC | H,R 76 | XP_005256679.1 | |
XM_006721515.2 | 294 | Intron | XP_006721578.1 |