Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGGATCTGGAACAGTACGCGG[C/T]CAGCTACAGCGGCCTGATGCGCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601934 MIM: 602578 | ||||||||||||||||||||
Literature Links: |
DUS1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUS1L - dihydrouridine synthase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPS1 - G protein pathway suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321089.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | NP_001308018.1 | |
NM_001321090.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | NP_001308019.1 | |
NM_001321091.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | NP_001308020.1 | |
NM_001321092.1 | 385 | Missense Mutation | GCC,GTC | A,V 49 | NP_001308021.1 | |
NM_001321093.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | NP_001308022.1 | |
NM_004127.5 | 385 | Missense Mutation | GCC,GTC | A,V 49 | NP_004118.3 | |
NM_212492.2 | 385 | Missense Mutation | GCC,GTC | A,V 89 | NP_997657.1 | |
XM_005256350.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | XP_005256407.1 | |
XM_005256353.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | XP_005256410.1 | |
XM_005256359.1 | 385 | Missense Mutation | GCC,GTC | A,V 49 | XP_005256416.1 | |
XM_005256360.3 | 385 | Missense Mutation | GCC,GTC | A,V 49 | XP_005256417.1 | |
XM_011523569.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_011521871.1 | |
XM_011523570.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_011521872.1 | |
XM_017024524.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880013.1 | |
XM_017024525.1 | 385 | Missense Mutation | GCC,GTC | A,V 117 | XP_016880014.1 | |
XM_017024526.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880015.1 | |
XM_017024527.1 | 385 | Missense Mutation | GCC,GTC | A,V 49 | XP_016880016.1 | |
XM_017024528.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880017.1 | |
XM_017024529.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880018.1 | |
XM_017024530.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880019.1 | |
XM_017024531.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880020.1 | |
XM_017024532.1 | 385 | Missense Mutation | GCC,GTC | A,V 49 | XP_016880021.1 | |
XM_017024533.1 | 385 | Missense Mutation | GCC,GTC | A,V 49 | XP_016880022.1 | |
XM_017024534.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880023.1 | |
XM_017024535.1 | 385 | Missense Mutation | GCC,GTC | A,V 33 | XP_016880024.1 |
RFNG - RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |