Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCATGTTCCAGTTTGAGCACAT[A/G]AAGTGGCAGTATGCCGTGGAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611679 | ||||||||||||||||||||
Literature Links: |
FBXW10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXW10 - F-box and WD repeat domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267585.1 | 2443 | Intron | NP_001254514.1 | |||
NM_001267586.1 | 2443 | Intron | NP_001254515.1 | |||
XM_011523625.1 | 2443 | Intron | XP_011521927.1 | |||
XM_011523626.2 | 2443 | Intron | XP_011521928.1 | |||
XM_017024020.1 | 2443 | Missense Mutation | ATA,ATG | I,M 737 | XP_016879509.1 | |
XM_017024021.1 | 2443 | Silent Mutation | TAA,TGA | *,* 725 | XP_016879510.1 |
TVP23B - trans-golgi network vesicle protein 23 homolog B (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |