Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTACCTTTGTAGATTTCTCTAA[C/T]GGTTGATCATACTGAGCCTGCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604694 | ||||||||||||||||||||
Literature Links: |
AKAP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP10 - A-kinase anchoring protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007202.3 | 2142 | Silent Mutation | CCA,CCG | P,P 655 | NP_009133.2 | |
XM_006721432.3 | 2142 | Silent Mutation | CCA,CCG | P,P 597 | XP_006721495.1 | |
XM_017024071.1 | 2142 | Intron | XP_016879560.1 |