Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTATGGTGCACCTGGCGTGGA[A/G]TTCACAGGCCTGCACCGGGATGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LLGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LLGL1 - LLGL1, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004140.3 | 262 | Silent Mutation | GAA,GAG | E,E 67 | NP_004131.3 | |
XM_011523849.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522151.1 | |
XM_011523850.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522152.1 | |
XM_011523851.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522153.1 | |
XM_011523852.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522154.1 | |
XM_011523853.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522155.1 | |
XM_011523854.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522156.1 | |
XM_011523855.2 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522157.1 | |
XM_011523856.1 | 262 | Silent Mutation | GAA,GAG | E,E 67 | XP_011522158.1 |