Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCTGTGAGGACCTCTTCTCTCG[A/C]GTTAGTGAGAACCAGAGTGCTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INCA1 - inhibitor of CDK, cyclin A1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167985.1 | 735 | Intron | NP_001161457.1 | |||
NM_001167986.1 | 735 | Intron | NP_001161458.1 | |||
NM_001167987.1 | 735 | Intron | NP_001161459.1 | |||
NM_213726.2 | 735 | Intron | NP_998891.2 |
KIF1C - kinesin family member 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006612.5 | 735 | Silent Mutation | CGA,CGC | R,R 129 | NP_006603.2 | |
XM_005256424.2 | 735 | Silent Mutation | CGA,CGC | R,R 129 | XP_005256481.1 |