Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCACATCTGTCCCCCCAGCAAAC[C/T]GCTGCAAGAAGGCCCCAGTGAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147557 | ||||||||||||||||||||
Literature Links: |
ITGB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGB4 - integrin subunit beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000213.4 | 296 | Missense Mutation | CGC,TGC | R,C 29 | NP_000204.3 | |
NM_001005619.1 | 296 | Missense Mutation | CGC,TGC | R,C 29 | NP_001005619.1 | |
NM_001005731.2 | 296 | Missense Mutation | CGC,TGC | R,C 29 | NP_001005731.1 | |
NM_001321123.1 | 296 | Missense Mutation | CGC,TGC | R,C 29 | NP_001308052.1 | |
XM_005257309.2 | 296 | Missense Mutation | CGC,TGC | R,C 29 | XP_005257366.1 | |
XM_005257311.4 | 296 | Missense Mutation | CGC,TGC | R,C 29 | XP_005257368.1 | |
XM_006721866.3 | 296 | Missense Mutation | CGC,TGC | R,C 64 | XP_006721929.1 | |
XM_006721867.3 | 296 | Missense Mutation | CGC,TGC | R,C 64 | XP_006721930.1 | |
XM_006721868.3 | 296 | Missense Mutation | CGC,TGC | R,C 64 | XP_006721931.1 | |
XM_006721870.3 | 296 | Missense Mutation | CGC,TGC | R,C 64 | XP_006721933.1 | |
XM_011524751.2 | 296 | Missense Mutation | CGC,TGC | R,C 64 | XP_011523053.1 | |
XM_011524752.2 | 296 | Intron | XP_011523054.1 |